Lanthanum(III) sulfate hydrate ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
43106-22
43106-36
CAAA43106-22EA
161.16
CAD
CAAA43106-22
CAAA43106-36
Lanthanum(III) sulfate hydrate ≥99.9% (REO, rare earth oxide basis)
Lanthanum (III) sulphate hydrate
MDL: MFCD00011070
Purity: 99.9%
CAS No.: 57804-25-8
Molecular Formula: La2(SO4)3.xH2O
Formula Weight: 566.00(anhy)
Physical Form: Crystalline
Density: 2.821
Melting Point: dec. white heat
Merck Reference: 14,5363
MDL No.: MFCD00011070
Formula:
La₂O₁₂S₃·H₂O Melting Pt: Dec. (white heat) Density: 2.821 Storage Temperature: Ambient |
MDL Number:
MFCD00011070 CAS Number: 57804-25-8 EINECS: 233-239-6 Merck Index: 14,5363 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...