Titanium ≥99.5% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

43102-18 43102-30
CAAA43102-18EA 74.1 CAD
CAAA43102-18 CAAA43102-30
Titanium ≥99.5% (metals basis), powder -200 mesh
Titanium

MDL: MFCD00011264


Purity: 99.5% CAS No.: 7440-32-6 Odor: Odorless UN No.: 2546 MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3277 °C (1013 hPa)
Melting Pt: 1677 °C
Density: 4,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR