Antimony (III) ethanolate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

43097-06 43097-14 43097-22
CAAA43097-06EA 131.93 CAD
CAAA43097-06 CAAA43097-14 CAAA43097-22
Antimony (III) ethanolate ≥99.9% (metals basis)
Antimony(III) ethanolate

MDL: MFCD00015123


Purity: 99.9% CAS No.: 10433-06-4 Molecular Formula: Sb(OC2H5)3 Formula Weight: 256.90 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.524 Sensitivity: Moisture Sensitive Boiling Point: 95°/11mm UN No.: 3082 MDL No.: MFCD00015123

Formula: C₆H₁₅O₃Sb
Boiling Pt: 95 °C (11 mmHg)
Density: 1,524
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015123
CAS Number: 10433-06-4
EINECS: 233-914-5
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR