Platinum(II) chloride ≥99.99% (metals basis) Pt ≥ 73%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Platinum dichloride
43090-03
43090-06
CAAA43090-03EA
486.64
CAD
CAAA43090-03
CAAA43090-06
Platinum(II) chloride ≥99.99% (metals basis) Pt ≥ 73%, Premion®
Platinum (II) chloride
MDL: MFCD00011181
Purity: 99.99+%
CAS No.: 10025-65-7
Molecular Formula: Cl2Pt
Formula Weight: 266.00
Physical Form: Powder
Density: 5.87
Appearance: Green-brown
Melting Point: 581° dec.
Merck Reference: 14,7528
UN No.: 3260
MDL No.: MFCD00011181
Formula:
Cl₂Pt MW: 265.99 g/mol Melting Pt: 581 (dec.) Density: 5.87 Storage Temperature: Ambient |
MDL Number:
MFCD00011181 CAS Number: 10025-65-7 EINECS: 233-034-1 UN: 3260 ADR: 8 III Merck Index: 14,7528 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...