Silver nitrate ≥99.995% (metals basis) Ag ≥ 63%, Premion®
Supplier: Thermo Scientific Chemicals
|
Danger
|
43087-06
43087-14
43087-22
CAAA43087-06EA
71.42
CAD
CAAA43087-06
CAAA43087-14
CAAA43087-22
Silver nitrate ≥99.995% (metals basis) Ag ≥ 63%, Premion®
Silver nitrate
MDL: MFCD00003414
Purity: 99.995%
CAS No.: 7761-88-8
Molecular Formula: AgNO3
Formula Weight: 169.87
Physical Form: Crystalline
Density: 4.352
Appearance: White
Odor: Odorless
Melting Point: 212°
Merck Reference: 14,8518
UN No.: 1479
MDL No.: MFCD00003414
Formula:
AgNO₃ Boiling Pt: 444 °C (1013 hPa) Melting Pt: 212 °C Density: 4.352 Storage Temperature: Ambient |
MDL Number:
MFCD00003414 CAS Number: 7761-88-8 EINECS: 231-853-9 UN: 1479 ADR: 5.1,I REACH: 01-2119513705-43 Merck Index: 14,8518 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...