Palladium(II) acetate trimer (≥47% Pd) ≥99.98% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43086-03
43086-06
CAAA43086-03EA
244.9
CAD
CAAA43086-03
CAAA43086-06
Palladium(II) acetate trimer (≥47% Pd) ≥99.98% (metals basis)
Palladium (II) acetate
MDL: MFCD00012453
Purity: 99.98%
CAS No.: 3375-31-3
Molecular Formula: C12H18O12Pd3
Formula Weight: 673.46
Physical Form: Powder
Appearance: Red-brown
Melting Point: ca 205° dec.
Merck Reference: 14,6991
MDL No.: MFCD00012453
Formula:
C₄H₆O₄Pd Melting Pt: ∼ 205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012453 CAS Number: 3375-31-3 EINECS: 222-164-4 Merck Index: 14,6991 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...