Palladium(II) chloride (≥59.5% Pd) ≥99.999% (metals basis) Pd ≥ 59.5%, Premion®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Palladium dichloride

43085-03 43085-06 43085-14
CAAA43085-03EA 314.42 CAD
CAAA43085-03 CAAA43085-06 CAAA43085-14
Palladium(II) chloride (≥59.5% Pd) ≥99.999% (metals basis) Pd ≥ 59.5%, Premion®
Palladium (II) chloride

MDL: MFCD00003558


Purity: 99.999% CAS No.: 7647-10-1 Molecular Formula: PdCl2 Formula Weight: 177.31 Physical Form: Powder Density: 4.0 Appearance: Red-brown Odor: Odorless Melting Point: 675° dec. Merck Reference: 14,6990 MDL No.: MFCD00003558

Formula: PdCl₂
MW: 177.33 g/mol
Melting Pt: 675 °C (dec.)
Density: 4.0
Storage Temperature: Ambient
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8 III
Merck Index: 14,6990

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR