Palladium(II) iodide ≥99.998% (metals basis) Pd ≥ 29%, Premion®
Supplier: Thermo Scientific Chemicals
|
Warning
|
43084-04
43084-09
CAAA43084-04EA
535.62
CAD
CAAA43084-04
CAAA43084-09
Palladium(II) iodide ≥99.998% (metals basis) Pd ≥ 29%, Premion®
Palladium (II) iodide
MDL: MFCD00011171
Purity: 99.998%
CAS No.: 7790-38-7
Molecular Formula: PdI2
Formula Weight: 360.23
Physical Form: Powder
Density: 6.003
Appearance: Black
Melting Point: 350° dec.
MDL No.: MFCD00011171
Formula:
I₂Pd Melting Pt: 350 °C (dec.) Density: 6.003 Storage Temperature: Ambient |
MDL Number:
MFCD00011171 CAS Number: 7790-38-7 EINECS: 232-203-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...