Niobium hard ≥99.8% (metals basis), rods, hard, Ø 19 mm (0.75 in)

Supplier: Thermo Scientific Chemicals

43080-BP 43080-BS
CAAA43080-BPEA 167.48 CAD
CAAA43080-BP CAAA43080-BS
Niobium hard ≥99.8% (metals basis), rods, hard, Ø 19 mm (0.75 in)
Niobium

MDL: MFCD00011126


Purity: 99.8% CAS No.: 7440-03-1 Physical Form: ≈24.3g/cm, Temper: as drawn Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR