Toluene 99,8, Environmental Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Methylbenzene, Phenylmethane, FM approval
Suitable for pesticide residue analysis
Purity: 99.8+%
CAS No.: 108-88-3
Molecular Formula: C6H5CH3
Formula Weight: 92.14
Flash Point: 4°(40°F)
Physical Form: Liquid
Density: 0.867
Refractive Index: 1.4967
Odor: Aromatic
Melting Point: -93°
Boiling Point: 111°
Beilstein Reference: 635760
Merck Reference: 14,9529
UN No.: 1294
MDL No.: MFCD00008512
Suitable for pesticide residue analysis
Formula:
C₆H₅CH₃ MW: 92.14 g/mol Boiling Pt: 110.6 Melting Pt: -95 Density: 0.867 Flash Pt: 4 °C (40 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008512 CAS Number: 108-88-3 EINECS: 203-625-9 UN: 1294 ADR: 3 II REACH: 05-2114615130-69 Merck Index: 14,9529 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...