Zirconium ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Scientific Chemicals

43049-FF 43049-FQ 43049-FW
CAAA43049-FFEA 109.34 CAD
CAAA43049-FF CAAA43049-FQ CAAA43049-FW
Zirconium ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Zirconium

MDL: MFCD00011303


Purity: 99.9+% CAS No.: 7440-67-7 Physical Form: ≈0.1g/25x25mm Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
MW: 91.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR