Cobalt ≥99.95% (metals basis), rods, Ø 5 mm (0.2 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
43032-DJ
43032-DO
CAAA43032-DJEA
237
CAD
CAAA43032-DJ
CAAA43032-DO
Cobalt ≥99.95% (metals basis), rods, Ø 5 mm (0.2 in)
Cobalt
MDL: MFCD00010935
Purity: 99.95%
CAS No.: 7440-48-4
Physical Form: ≈1.75g/cm
Odor: Odorless
MDL No.: MFCD00010935
Formula:
Co MW: 58.93 g/mol Boiling Pt: 2870 Melting Pt: 1495 Density: 8.9 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 Merck Index: 12,02488 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...