Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 19 mm (0.75 in)

Supplier: Thermo Scientific Chemicals

43031-DH 43031-HF
CAAA43031-DHEA 1403.04 CAD
CAAA43031-DH CAAA43031-HF
Gadolinium ≥99.9% (metals basis excluding Ta), rods, Ø 19 mm (0.75 in)
Gadolinium

MDL: MFCD00011022


Purity: 99.9% CAS No.: 7440-54-2 Molecular Formula: Packaged in mylar under argon Physical Form: ≈2.2g/mm, Cast rod, diameter is approximate MDL No.: MFCD00011022

Formula: Gd
Storage Temperature: Ambient
MDL Number: MFCD00011022
CAS Number: 7440-54-2
EINECS: 231-162-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR