Tetraethylammonium hydroxide 35% (w/w) in aqueous solution, Reagent Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
43023-14
43023-22
43023-36
CAAA43023-14EA
51.19
CAD
CAAA43023-14
CAAA43023-22
CAAA43023-36
Tetraethylammonium hydroxide 35% (w/w) in aqueous solution, Reagent Grade
Tetraethylammonium hydroxide
MDL: MFCD00009024
Beilstein Registry No.: 3912961
Notes: 10.1% halide, Na and K less than 20ppm
CAS No.: 77-98-5
Molecular Formula: C8H21NO
Formula Weight: 147.26
Physical Form: Liquid
Density: 1.023
Refractive Index: 1.4040
Sensitivity: Air Sensitive
Odor: Amine-like
Boiling Point: dec.
Beilstein Reference: 3912961
Merck Reference: 14,9201
UN No.: 3267
MDL No.: MFCD00009024
Formula:
(C₂H₅)₄N(OH) Boiling Pt: Dec. Density: 1.023 Storage Temperature: Ambient |
MDL Number:
MFCD00009024 CAS Number: 77-98-5 EINECS: 201-073-3 UN: 3267 ADR: 8,II Merck Index: 14,9201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...