Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), wire, annealed, Ø 0.127 mm (0.005 in)
Supplier: Thermo Scientific Chemicals
43019-BS
43019-BW
CAAA43019-BSEA
87.06
CAD
CAAA43019-BS
CAAA43019-BW
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), wire, annealed, Ø 0.127 mm (0.005 in)
Platinum Iridium alloy (90:10 w%)
Applications: Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.03g/10cm, Temper: soft
Odor: Odorless
MDL No.: MFCD02091683
Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...