Palladium ≥99.9% (metals basis), powder , -60 mesh
Supplier: Thermo Scientific Chemicals
|
Warning
|
43010-04
43010-09
CAAA43010-04EA
1044.38
CAD
CAAA43010-04
CAAA43010-09
Palladium ≥99.9% (metals basis), powder , -60 mesh
Palladium
MDL: MFCD00011167
Purity: 99.9%
CAS No.: 7440-05-3
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011167
Formula:
Pd Density: 12.02 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011167 CAS Number: 7440-05-3 EINECS: 231-115-6 UN: 3089 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...