Silicon ≥99.9999% (metals basis), lumps 0.1-2.5cm (0.04-1.0in)

Supplier: Thermo Scientific Chemicals
Warning

43006-22 43006-36
CAAA43006-22EA 276.5 CAD
CAAA43006-22 CAAA43006-36
Silicon ≥99.9999% (metals basis), lumps 0.1-2.5cm (0.04-1.0in)
Silicon

MDL: MFCD00085311


Purity: 99.9999% CAS No.: 7440-21-3 Odor: Odorless MDL No.: MFCD00085311

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR