Cesium ≥99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

43005-03 43005-06 43005-18
CAAA43005-03EA 156.42 CAD
CAAA43005-03 CAAA43005-06 CAAA43005-18
Cesium ≥99.8% (metals basis)
Cesium

MDL: MFCD00134037 Purity based on metal contaminates only. Vacuum sealed in ampoules.


Purity: 99.8% CAS No.: 7440-46-2 Physical Form: Ampouled Odor: Odorless UN No.: 1407 MDL No.: MFCD00134037

Formula: Cs
Boiling Pt: 705 °C (1013 hPa)
Melting Pt: 28.5 °C
Density: 1.873 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134037
CAS Number: 7440-46-2
EINECS: 231-155-4
UN: 1407
ADR: 4.3,I
Merck Index: 12,02051

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR