Silver ≥99.999% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42984-06 42984-14 42984-22
CAAA42984-06EA 152.63 CAD
CAAA42984-06 CAAA42984-14 CAAA42984-22
Silver ≥99.999% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Silver

MDL: MFCD00003397


Purity: 99.999% CAS No.: 7440-22-4 Physical Form: ≈0.26g/slug Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR