Gold germanium alloy (88:12 w%) ≥99.998% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42983-KJ 42983-KM
CAAA42983-KJEA 796.32 CAD
CAAA42983-KJ CAAA42983-KM
Gold germanium alloy (88:12 w%) ≥99.998% (metals basis), slug, Premion®, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Gold Germanium alloy (88:12 w%)

MDL: MFCD02091706


Purity: 99.998% Molecular Formula: Au:Ge; 88:12 wt% Physical Form: ≈0.4g/slug Density: 0.37G/CM(3) Odor: Odorless MDL No.: MFCD02091706

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR