Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.5 mm (0.02 in)
Supplier: Thermo Scientific Chemicals
42982-BU
42982-BY
42982-CC
CAAA42982-BUEA
71.42
CAD
CAAA42982-BU
CAAA42982-BY
CAAA42982-CC
Hafnium ≥99.95% (metals basis excluding Zr), wire (Zr ∼ 3%), Ø 0.5 mm (0.02 in)
Hafnium
MDL: MFCD00011032
Purity: 99.95%
CAS No.: 7440-58-6
Physical Form: ≈2.6g/m
Odor: Odorless
MDL No.: MFCD00011032
Formula:
Hf MW: 178.49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011032 CAS Number: 7440-58-6 EINECS: 231-166-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...