Copper ≥99.9999% (metals basis), foil, Puratronic®, Thickness: 0.05 mm (0.002 in)
Supplier: Thermo Scientific Chemicals
42972-FI
42972-GH
CAAA42972-FIEA
158
CAD
CAAA42972-FI
CAAA42972-GH
Copper ≥99.9999% (metals basis), foil, Puratronic®, Thickness: 0.05 mm (0.002 in)
Copper
MDL: MFCD00010965
Purity: 99.9999%
CAS No.: 7440-50-8
Physical Form: ≈1.1g/50x50mm
Odor: Odorless
MDL No.: MFCD00010965
Formula:
Cu Boiling Pt: 2595 °C (1013 hPa) Melting Pt: 1083 °C Density: 8.94 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 Merck Index: 13,02545 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...