Borane dimethyl sulfide complex 2 M in THF, chemSeal™
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Borane-methyl sulfide complex, Borane-methyl sulphide complex
Packaged under argon in resealable ChemSeal™ bottles.
42965-AE
42965-AP
CAAA42965-AEEA
124.82
CAD
CAAA42965-AE
CAAA42965-AP
Borane dimethyl sulfide complex 2 M in THF, chemSeal™
Borane dimethyl sulphide complex
MDL: MFCD00013189
Beilstein Registry No.: 3663489
CAS No.: 13292-87-0
Molecular Formula: C2H9BS
Formula Weight: 75.97
Flash Point: -17°(-1°F)
Physical Form: Liquid
Density: 0.855
Appearance: Colorless to pale yellow
Odor: Characteristic
Beilstein Reference: 3663489
UN No.: 3399
MDL No.: MFCD00013189
Formula:
C₂H₉BS Density: 0.855 Flash Pt: –17 °C (1 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013189 CAS Number: 13292-87-0 EINECS: 236-313-6 UN: 3399 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...