Selenium disulfide 94%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Selenium(IV) sulfide, Selenium sulphide, Selenium(IV) sulphide, Selenium sulfide

42943-14 42943-22 42943-36
CAAA42943-14EA 76.16 CAD
CAAA42943-14 CAAA42943-22 CAAA42943-36
Selenium disulfide 94%
Selenium disulphide

MDL: MFCD00011227 Insoluble in water, in dilute HCl


Purity: 94% CAS No.: 7488-56-4 Molecular Formula: SeS2 Formula Weight: 143.09 Physical Form: Powder Melting Point: 100° Boiling Point: dec. Solubility: Insoluble in water, in dilute HCl Merck Reference: 14,8436 UN No.: 2657 MDL No.: MFCD00011227

Formula: S₂Se
Boiling Pt: Dec.
Melting Pt: <100 °C
Density: 0.03 g/cm³ (–65 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011227
CAS Number: 7488-56-4
EINECS: 231-303-8
UN: 2657
ADR: 6.1,II
Merck Index: 14,8436

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR