Titanium(II) oxide ≥99.9% (metals basis), pellets
Supplier: Thermo Scientific Chemicals
Synonyms:
Titanium oxide, Titanium monoxide
42940-14
42940-22
CAAA42940-14EA
202.24
CAD
CAAA42940-14
CAAA42940-22
Titanium(II) oxide ≥99.9% (metals basis), pellets
Titanium (II) oxide
MDL: MFCD00036279
Notes: For vacuum deposition
Purity: 99.9%
CAS No.: 12137-20-1
Molecular Formula: TiO
Formula Weight: 63.90
Physical Form: 8-9mm(0.32-0.36in) dia x 6-7mm(0.24-0.28in) thick
Density: 4.95
Melting Point: 1700°
Boiling Point:
Formula:
OTi MW: 63.88 Boiling Pt: 3660 Melting Pt: 1750 Density: 4.95 Storage Temperature: Ambient |
MDL Number:
MFCD00036279 CAS Number: 12137-20-1 EINECS: 235-236-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...