Titanium(II) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Titanium oxide, Titanium monoxide

42939-14 42939-22 42939-36
CAAA42939-14EA 176.96 CAD
CAAA42939-14 CAAA42939-22 CAAA42939-36
Titanium(II) oxide ≥99.9% (metals basis)
Titanium (II) oxide

MDL: MFCD00036279


Purity: 99.9% CAS No.: 12137-20-1 Molecular Formula: TiO Formula Weight: 63.90 Physical Form: Lump Density: 4.95 Melting Point: 1700° Boiling Point: 3000° MDL No.: MFCD00036279

Formula: OTi
MW: 63.88
Boiling Pt: >3000 °C
Melting Pt: 1700 °C
Density: 4.88
Storage Temperature: Ambient
MDL Number: MFCD00036279
CAS Number: 12137-20-1
EINECS: 235-236-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR