Chromium 99,999 Metals basis, granules -4/+7 mesh

Supplier: Thermo Scientific Chemicals

42926-18 42926-30
CAAA42926-18EA 461.36 CAD
CAAA42926-18 CAAA42926-30
Chromium 99,999 Metals basis, granules -4/+7 mesh
Chromium

MDL: MFCD00010944


Purity: 99.999% CAS No.: 7440-47-3 Odor: Odorless MDL No.: MFCD00010944

Formula: Cr
MW: 52
Boiling Pt: 2469
Melting Pt: 1840
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR