Copper(II) sulfide ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Copper sulfide, Cupric sulfide, Copper sulphide, Cupric sulphide, Copper(II)sulfide, Copper(II)sulphide
42925-18
42925-30
42925-A1
CAAA42925-18EA
132.4
CAD
CAAA42925-18
CAAA42925-30
CAAA42925-A1
Copper(II) sulfide ≥99.8% (metals basis)
Copper (II) sulphide
MDL: MFCD00016066
Purity: 99.8%
CAS No.: 1317-40-4
Molecular Formula: CuS
Formula Weight: 95.60
Physical Form: -200 Mesh Powder
Density: 3.9-4.6
Refractive Index: 1.45
Sensitivity: Air and Moisture Sensitive
Melting Point: 220° dec.
Merck Reference: 14,2655
MDL No.: MFCD00016066
Formula:
CuS MW: 95.61 g/mol Melting Pt: 220 °C (dec.) Density: 3.9…4.6 Storage Temperature: Ambient |
MDL Number:
MFCD00016066 CAS Number: 1317-40-4 EINECS: 215-271-2 Merck Index: 14,2655 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...