Silver ≥99.99% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42924-06 42924-14
CAAA42924-06EA 96.38 CAD
CAAA42924-06 CAAA42924-14
Silver ≥99.99% (metals basis), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Silver

MDL: MFCD00003397


Purity: 99.99% CAS No.: 7440-22-4 Physical Form: ≈0.26g/slug Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212 °C (1013 hPa)
Melting Pt: 962 °C
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR