Aluminium ≥99.97% (metals basis), powder -325 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
42920-06
42920-22
CAAA42920-06EA
60.36
CAD
CAAA42920-06
CAAA42920-22
Aluminium ≥99.97% (metals basis), powder -325 mesh
Aluminium
MDL: MFCD00134029
Purity: 99.97%
CAS No.: 7429-90-5
Physical Form: APS approximately 15 micron
Sensitivity: Moisture Sensitive
Appearance: Silver-white-grey
Odor: Odorless
UN No.: 1396
MDL No.: MFCD00134029
Formula:
Al Boiling Pt: 2057 °C (1013 hPa) Melting Pt: 659.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 UN: 1396 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...