1,3-Dioxolane ≥99.5% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
42897-AK
42897-K2
42897-M1
CAAA42897-AKEA
67.15
CAD
CAAA42897-AK
CAAA42897-K2
CAAA42897-M1
1,3-Dioxolane ≥99.5% stabilized
1,3-Dioxolane
As a solvent, in Mannich reaction, as an electrolyte in batteries
Purity: 99.5%
CAS No.: 646-06-0
Molecular Formula: C3H6O2
Formula Weight: 74.08
Flash Point: 1°(34°F)
Physical Form: Liquid
Density: 1.06
Refractive Index: 1.4000
Melting Point: -95°
Boiling Point: 74-75°
UN No.: 1166
MDL No.: MFCD00003207
As a solvent, in Mannich reaction, as an electrolyte in batteries
Formula:
C₃H₆O₂ MW: 74.08 g/mol Boiling Pt: 74…75 °C Melting Pt: –95 °C Density: 1.06 Flash Pt: 1 °C (34 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003207 CAS Number: 646-06-0 EINECS: 211-463-5 UN: 1166 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...