Titanium ≥99.7% (metals basis), plate 100x100 mm (3.9x3.9 in), annealed, Thickness: 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals
Warning

42896-KG 42896-KJ
CAAA42896-KGEA 344.44 CAD
CAAA42896-KG CAAA42896-KJ
Titanium ≥99.7% (metals basis), plate 100x100 mm (3.9x3.9 in), annealed, Thickness: 6.35 mm (0.25 in)
Titanium

MDL: MFCD00011264


Purity: 99.7% CAS No.: 7440-32-6 Physical Form: ≈286g/pc Odor: Odorless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR