Europium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid europium (III) salt hydrate, Europium(III) acetate xhydrate, Europium (III) acetate hydrate

42878-06 42878-14
CAAA42878-06EA 178.54 CAD
CAAA42878-06 CAAA42878-14
Europium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Europium triacetate hydrate

MDL: MFCD00150115


Purity: 99.9% CAS No.: 62667-64-5 Molecular Formula: Eu(OOCCH3)3.xH2O Formula Weight: 329.10(anhy) Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00150115

Formula: C₆H₉EuO₆·H₂O
MW: 347.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150115
CAS Number: 62667-64-5
EINECS: 214-670-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR