Platinum 99,9 Metals basis, sponge ≤ 2 mm (0.08 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Platinium

42871-03 42871-06
CAAA42871-03EA 497.7 CAD
CAAA42871-03 CAAA42871-06
Platinum 99,9 Metals basis, sponge ≤ 2 mm (0.08 in)
Platinum

MDL: MFCD00011179


Purity: 99.9% CAS No.: 7440-06-4 Odor: Odorless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR