Diyttrium trioxide ≥99.9999% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Scientific Chemicals
|
Warning
|
42864-09
42864-18
42864-30
CAAA42864-09EA
173.8
CAD
CAAA42864-09
CAAA42864-18
CAAA42864-30
Diyttrium trioxide ≥99.9999% (REO, rare earth oxide basis), REacton®
Diyttrium trioxide
MDL: MFCD00011473
Purity: 99.9999%
CAS No.: 1314-36-9
Molecular Formula: Y2O3
Formula Weight: 225.81
Physical Form: Powder
Density: 5.01
Appearance: White
Odor: Odorless
Melting Point: 2410°
Merck Reference: 14,10107
MDL No.: MFCD00011473
Formula:
Y₂O₃ Boiling Pt: 4300 °C (1013 hPa) Melting Pt: 2410 °C Density: 5.01 Storage Temperature: Ambient |
MDL Number:
MFCD00011473 CAS Number: 1314-36-9 EINECS: 215-233-5 Merck Index: 14,10107 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...