Bismuth(III) oxide ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

42863-18 42863-30 42863-A1
CAAA42863-18EA 188.02 CAD
CAAA42863-18 CAAA42863-30 CAAA42863-A1
Bismuth(III) oxide ≥99.999% (metals basis), Puratronic®
Bismuth (III) oxide

MDL: MFCD00003462


Purity: 99.999% CAS No.: 1304-76-3 Molecular Formula: Bi2O3 Formula Weight: 465.96 Physical Form: Powder Density: 8.9 Appearance: Yellow Melting Point: 817° Boiling Point: ca 1890° Merck Reference: 14,1273 MDL No.: MFCD00003462

Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890
Melting Pt: 825
Density: 8.9
Storage Temperature: Ambient
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 14,1273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR