Silver ≥99.9% (metals basis), wool

Supplier: Thermo Scientific Chemicals

42860-03 42860-06 42860-14
CAAA42860-03EA 92.75 CAD
CAAA42860-03 CAAA42860-06 CAAA42860-14
Silver ≥99.9% (metals basis), wool
Silver

MDL: MFCD00003397


Purity: 99.9+% CAS No.: 7440-22-4 Physical Form: Approximately 0.05mm dia fibers Odor: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR