Magnesium chloride, ultra dry ≥99.99% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

42850-06 42850-14 42850-22
CAAA42850-06EA 89.9 CAD
CAAA42850-06 CAAA42850-14 CAAA42850-22
Magnesium chloride, ultra dry ≥99.99% (metals basis)
Magnesium chloride

MDL: MFCD00011106


Purity: 99.99% CAS No.: 7786-30-3 Molecular Formula: Cl2Mg Formula Weight: 95.22 Physical Form: Powder, Ampouled under argon Density: 2.32 Appearance: White to yellow Odor: Odorless Melting Point: 714° Merck Reference: 14,5662 MDL No.: MFCD00011106

Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 715
Density: 2.32
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 14,5662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR