Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

42848-14 42848-22
CAAA42848-14EA 99.54 CAD
CAAA42848-14 CAAA42848-22
Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Niobium

MDL: MFCD00011126


Purity: 99.95% CAS No.: 7440-03-1 Physical Form: ≈3.4g/slug Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR