Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)

Supplier: Thermo Scientific Chemicals

42847-14 42847-22
CAAA42847-14EA 108.23 CAD
CAAA42847-14 CAAA42847-22
Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Niobium

MDL: MFCD00011126


Purity: 99.95% CAS No.: 7440-03-1 Physical Form: ≈1.7g/slug Odor: Odorless MDL No.: MFCD00011126

Formula: Nb
MW: 92.91 g/mol
Boiling Pt: 3700
Melting Pt: 2468
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR