Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
42846-09
42846-18
CAAA42846-09EA
73.79
CAD
CAAA42846-09
CAAA42846-18
Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Niobium
MDL: MFCD00011126
Purity: 99.95%
CAS No.: 7440-03-1
Physical Form: ≈0.43g/slug
Odor: Odorless
MDL No.: MFCD00011126
Formula:
Nb Boiling Pt: 3700 °C (1013 hPa) Melting Pt: 2468 °C Density: 8.57 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011126 CAS Number: 7440-03-1 EINECS: 231-113-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...