Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42845-09 42845-18
CAAA42845-09EA 95.43 CAD
CAAA42845-09 CAAA42845-18
Niobium ≥99.95% (metals basis excluding Ta), slug, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Niobium

For vapor deposition


Purity: 99.95% CAS No.: 7440-03-1 Physical Form: ≈0.22g/slug Odor: Odorless MDL No.: MFCD00011126


For vapor deposition

Formula: Nb
Boiling Pt: 3700 °C (1013 hPa)
Melting Pt: 2468 °C
Density: 8.57 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011126
CAS Number: 7440-03-1
EINECS: 231-113-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR