Lithium chloride, ultra dry ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

42842-06 42842-14 42842-22
CAAA42842-06EA 67.94 CAD
CAAA42842-06 CAAA42842-14 CAAA42842-22
Lithium chloride, ultra dry ≥99.9% (metals basis)
Lithium chloride

MDL: MFCD00011078 Fieser: 1,609 2,246 4,298 5,677 13,332 16,194 18,211 20,215 21,248


Purity: 99.9% CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: Crystalline, Ampouled under argon Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078

Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2.068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR