Nickel ≥99.999% (metals basis), rods, Puratronic®, Ø 5 mm (0.2 in)

Supplier: Thermo Scientific Chemicals
Danger

42835-DJ 42835-DO
CAAA42835-DJEA 451.88 CAD
CAAA42835-DJ CAAA42835-DO
Nickel ≥99.999% (metals basis), rods, Puratronic®, Ø 5 mm (0.2 in)
Nickel

MDL: MFCD00011137


Purity: 99.999% CAS No.: 7440-02-0 Physical Form: ≈1.75g/cm Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
Boiling Pt: 3007 °C (1013 hPa)
Melting Pt: 1453 °C
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1,III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR