Vanadium ≥99.8% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)

Supplier: Thermo Scientific Chemicals

42829-06 42829-14
CAAA42829-06EA 128.45 CAD
CAAA42829-06 CAAA42829-14
Vanadium ≥99.8% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Vanadium

MDL: MFCD00011453


Purity: 99.8% CAS No.: 7440-62-2 Physical Form: ≈0.31g/slug Odor: Odorless MDL No.: MFCD00011453

Formula: V
MW: 50.94 g/mol
Boiling Pt: 3477
Melting Pt: 1896
Density: 6.015 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011453
CAS Number: 7440-62-2
EINECS: 231-171-1
Merck Index: 12,10053

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR