Tantalum ≥99.95% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Supplier: Thermo Scientific Chemicals
42827-09
42827-18
CAAA42827-09EA
159.58
CAD
CAAA42827-09
CAAA42827-18
Tantalum ≥99.95% (metals basis), slug, Length: 6.35 mm (0.25 in), Ø 3.175 mm (0.125 in)
Tantalum
MDL: MFCD00011252
Purity: 99.95%
CAS No.: 7440-25-7
Physical Form: ≈0.83g/slug
Odor: Odorless
MDL No.: MFCD00011252
Formula:
Ta Boiling Pt: 5427 °C (1013 hPa) Melting Pt: 2996 °C Density: 16.69 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011252 CAS Number: 7440-25-7 EINECS: 231-135-5 Merck Index: 12,09223 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...