Tungsten ≥99.95% (metals basis), wire, Ø 0.2 mm (0.008 in)
Supplier: Thermo Scientific Chemicals
42807-H2
42807-H7
CAAA42807-H2EA
274.92
CAD
CAAA42807-H2
CAAA42807-H7
Tungsten ≥99.95% (metals basis), wire, Ø 0.2 mm (0.008 in)
Tungsten
MDL: MFCD00011461
Purity: 99.95%
CAS No.: 7440-33-7
Physical Form: ≈0.61g/m
MDL No.: MFCD00011461
Formula:
W Boiling Pt: 5660 °C (1013 hPa) Melting Pt: 3410 °C Density: 19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011461 CAS Number: 7440-33-7 EINECS: 231-143-9 Merck Index: 12,09945 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...