tetra-Chloroauric(III) acid hydrate ≥99.999% (metals basis) Au ≥ 49.0%, Premion®

Supplier: Thermo Scientific Chemicals
Danger

42803-03 42803-06
CAAA42803-03EA 284.4 CAD
CAAA42803-03 CAAA42803-06
tetra-Chloroauric(III) acid hydrate ≥99.999% (metals basis) Au ≥ 49.0%, Premion®
tetra-Chloroauric (III) acid hydrate

MDL: MFCD00149903

Formula: HAuCl₄·H₂O
Density: 1.169 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149903
CAS Number: 27988-77-8
EINECS: 240-948-4
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR