Silicon(IV) oxide, amorphous, REacton® S.A. 105-130 m²/g, fumed, surface treated S.A. 105-130 m²/g

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silica gel amorphous, Silicon dioxide amorphous, Silicium dioxide amorphous

42784-18 42784-30 42784-A1
CAAA42784-18EA 31.13 CAD
CAAA42784-18 CAAA42784-30 CAAA42784-A1
Silicon(IV) oxide, amorphous, REacton® S.A. 105-130 m²/g, fumed, surface treated S.A. 105-130 m²/g
Silica gel amorphous

Coatings, adhesives, sealants, silicones, inks


CAS No.: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.09 Physical Form: -325 Mesh powder Density: 1.8 Appearance: White Odor: Odorless MDL No.: MFCD00011232


Coatings, adhesives, sealants, silicones, inks

Formula: SiO₂
MW: 60.08 g/mol
Melting Pt: 2230
Density: 1.8
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 7631-86-9
EINECS: 231-545-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR