Silicon(IV) oxide, amorphous, REacton® S.A. 105-145 m²/g, fumed, surface treated S.A. 105-145 m²/g

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Silica gel amorphous, Silicon dioxide amorphous, Silicium dioxide amorphous

42783-18 42783-30 42783-A1
CAAA42783-18EA 31.13 CAD
CAAA42783-18 CAAA42783-30 CAAA42783-A1
Silicon(IV) oxide, amorphous, REacton® S.A. 105-145 m²/g, fumed, surface treated S.A. 105-145 m²/g
Silica gel amorphous

Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity


CAS No.: 7631-86-9 Molecular Formula: O2Si Formula Weight: 60.09 Physical Form: -325 mesh powder Density: 2.2 Appearance: White Odor: Odorless MDL No.: MFCD00011232


Adhesives, coatings, sealants, inks, thermal insulation, silicones to provide hydrophobicity

Formula: SiO₂
MW: 60.08 g/mol
Melting Pt: ~ 2230 °C
Density: 2.2
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR